missing translation for 'onlineSavingsMsg'
Learn More
TRIS, 0.5M buffer soln., pH 7.4, Thermo Scientific™ Product Code.: 15455079

TRIS, 0.5M buffer soln., pH 7.4, Thermo Scientific™

Product Code. 15455079
500 mL, 500mL
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 15455079

Brand: Thermo Scientific Alfa Aesar J62778.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 0.5 M
pH 7.4
Physical Form Liquid
Quantity 500 mL
Solubility Information Not miscible or difficult to mix in water. Soluble in ethylene glycol (79.1mg/ml),methanol (26mg/ml),anhydrous ethanol (14.6mg/ml),95% ethanol (22mg/ml),DMF (14mg/ml),acetone (2mg/ml),ethyl acetate (0.5mg/ml),olive oil (0.4mg/ml),chloroform (0.05mg/ml),and alkaline solution.
Chemical Name or Material TRIS buffer soln.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TRIS, 0.5M buffer soln., pH 7.4, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.