missing translation for 'onlineSavingsMsg'
Learn More

Tris Buffered Saline Tablets, pH 7.6, Medicago

Supplied as exactly pre-weighed tablets. Medicago Tris Buffered Saline Tablets, pH 7.6 gives 500ml of 1x solution with 0.05M Tris-HCl buffer and 0.15M sodium chloride. Manufactured in ISO 9001 and 13485 certified facilities.

Brand:  Medicago 09-7500-100


Product Code. 11300499

  • 3830.00 DKK / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

TBS is isotonic and non-toxic to cells and is suitable for molecular biology. The buffer is commonly used as substance diluent or as wash buffer in immunoassays such as ELISA. It is used in immuno-histochemical staining when the background is high and for diluting alkaline phosphatase or peroxidase-conjugated antibodies in Western blotting.

  • Formulated from analytical grade reagents
  • Isotonic, non-toxic buffer
  • Guaranteed reproducibility
  • Ready to use in minutes
Specifications

Specifications

TBS
Wash buffer in immunoassays, antibody diluent in western blotting, sample diluent in in vitro diagnostics, immuno-histochemical staining, and in situ hybridization
Tablet
Tris-HCl (0.050M), NaKCl (0.15M)
White
7.6
100 x 500 mL Tablets
Analytical
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tris-Buffered Saline (TBS) pH8.0/pH7.6

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : Tris

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.