Learn More
HUBER Trolleys
Brand:  HUBER 6263
 View more versions of this product
Product Code. 10587695
  • 2535.00 DKK / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Specifications
Trolley
For Unistat 705, 705 W, 410 W
Product Title
HUBER Trolleys > For Use With: Unistat 705, 705w, 410w