Learn More
Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis), Thermo Scientific Chemicals
Evaporation source, contacts and coatings
388.41 DKK - 1297.20 DKK
Specifications
Chemical Name or Material | Tungsten wire, chromium plated |
---|---|
Assay Percent Notes | W ≥96.5%, Cr 99.888% (metals basis) |
MDL Number | MFCD00011461 |
Form | Wire |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319457
|
Thermo Scientific Alfa Aesar
041040.BS |
10 cm |
388.41 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15413765
|
Thermo Scientific Alfa Aesar
041040.8G |
5 x 10 cm |
794.81 DKK
10cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15423765
|
Thermo Scientific Alfa Aesar
041040.D@ |
10 x 10 cm |
1297.20 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Evaporation source, contacts and coatings
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tungsten wire, chromium plated | |
W ≥96.5%, Cr 99.888% (metals basis) | |
MFCD00011461 | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures