Learn More
WTW™ Turb™ Lamp Module
Brand:  WTW™ 600603
Product Code. 10488942
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spare Lamp Module
Turb 550 and Turb 555 Turbidimeter
Tungsten
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Turb™ Lamp Module > Tungsten