missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas TV200Y Standard Twin-Plate Wide Format Mini-Gel Electrophoresis Unit
Eliminates the problems associated with comparing samples from two separate tape-cast mini-gels and allows more samples to be screened on the same gel.
Brand: Scie Plas TV200Y
This item is not returnable.
View return policy
Description
- Casting base is included as standard
- Twin-plate 20.5W x 10cmH wide format mini-gel unit
- Designed specifically for PAGE techniques requiring self-cast gels
- Wide format of 20.5cm gel width effectively allows two mini-gels to be run within the same gel
- Glass plate stops within the newly modified GRM locks the glass plates firmly into position onto soft silicone gaskets in the casting base to ensure leak-free gel-casting
- Float glass plates of 4mm thick provides uniform gel thickness, sample migration and ground edges inhibits sample leakage during casting
- Twin screws on each gel-clamp acts in conjunction with a counterbalanced, dovetailed silicone gasket to disperse pressure evenly along the height and breadth of the gel as the screws are tightened
Specifications
1.5 to 2 hr. | |
W x H: 16.5 x 8.5 cm | |
Plate: 0.4 cm | |
Gel Tank: 2800 mL | |
W x H: 30 x 14 cm | |
100 to 150 V |
TV200Y Standard Twin-Plate Wide Format Mini-Gel Electrophoresis Unit | |
Twin-plate wide format mini-gel unit with GRM and gel tank, lid, 2 x (20.5W x 10cmH) plain glass plates, 4 x 1mm spacers, 2 x spacer aligners, 2 x (20.5 x 10cm) notched glass plates, 1 x dummy plate, 2 x 1mm thick 24-sample combs, casting base and 2 x silicone seals | |
0.1 cm | |
Inner Buffer Chamber: 300 mL | |
15 cm | |
10 to 15 mA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction