missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TWEAK (TNFSF12) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMTNFSF12X10
This item is not returnable.
View return policy
Description
TNF-related weak inducer of apoptosis (TWEAK) is a member of the tumor necrosis factor superfamily (TNFSF) of structurally related cytokines. Like most other members of this family, TWEAK is a cell surface-associated type II transmembrane protein although a smaller, biologically active form can also be generated by cleavage near the cell membrane. TWEAK has multiple biological activities, including stimulation of cell growth and angiogenesis, induction of inflammatory cytokines, in addition to stimulation of apoptosis. The TWEAK signal transduction pathway has not been well established but it appears to signal via TweakR/Fn14 in a manner similar to that described for other TNFSF members that bind receptors lacking death domains.Specifications
O54907 | |
0.3 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Tnfsf12 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
0.27 to 200 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
21944 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
TWEAK | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction