Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
Shop All Dupont Products
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
Description
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | Small |
Color | White |
Closure Type | Snaps |
Product Content Correction
Product Title
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck >
Spot an opportunity for improvement?Share a Content Correction