Description
- Not clean-processed and not sterilized
- Bound ties
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | Medium |
Color | White |
Cuff Style | Knit |
Model | IC702S00 |
Seam Type | Serged |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
48Hours
0Minutes
37Seconds
56New Offer Save Up to 92%
Product Content Correction
Product Title
DuPont™ Tyvek™ IsoClean™ Gown >
Spot an opportunity for improvement?Share a Content Correction