Description
- Shin length
- With neck and waist ties
- Antistatic treatment
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Specifications
Size | Universal |
Material | Polyethylene |
Color | White |
Construction | Flash-spun HDPE |
Includes | Neck and waist ties |
Certifications/Compliance | PPE Category 1, EN 1149-5 |
Item Description | Shin length apron with ties |
Seam Type | Seamless |
Documents
Scientific Resources
Flyers
DPT007 Tyvek accessories flyer UK
File size: 515.8 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
DuPont™ Tyvek 500 Apron >
Spot an opportunity for improvement?Share a Content Correction