Learn More
Labcon U-Pette™ Blood Transfer Dispensers
These blood transfer dispensers are for use with point of care testing instruments.
Brand: Labcon 7626-500-000-9
5410.35 DKK valid until 2025-02-28
Use promo code "24245" to get your promotional price.
This item is not returnable.
View return policy
Description
U-Pette™ Blood Dispenser for Point of Care Test Cartridges dispenses blood drops easily with no need to open collection tubes.
- Dispenses one or more drops of blood
- Fits standard glass or plastic blood tubes with rubber stoppers
- Includes Stainless Steel Cannula
- Not Autoclavable
Specifications
Blood Droplet Dispenser | |
Single-use | |
Point of Care Testing Instruments | |
500 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
CE_17404372.pdf
File size: 445.5 kB
CE Certificate
File size: 445.5 kB
Special Offers
-
Multiple Manufacturers
Life Sciences Monthly Promotion
Discover exclusive monthly deals on life sciences products to advance your research and innovation.Promo Code: 24245Expires: 2025-02-28
New Offer Save Up to 44%
Product Content Correction
Product Title
Labcon U-Pette™ Blood Transfer Dispensers > Blood Transfer Dispenser; Product Type: Blood Droplet Dispenser; For Use With (Equipment): Point of Care Testing Instruments; Disposable: Yes; Quantity: 500 Pk.
Spot an opportunity for improvement?Share a Content Correction