Learn More
ULBP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13133-1-AP-150UL
 View more versions of this product
Product Code. 16811204
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
ULBP2 encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells. The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety, or secreted. Many maligt cells secrete the encoded protein to evade immunosurveillance by NK cells. This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6.
Specifications
Specifications
ULBP2
Polyclonal
Unconjugated
ULBP2
ALCAN alpha, N2DL 2, N2DL2, NKG2D ligand 2, NKG2DL2, RAET1H, UL16 binding protein 2, ULBP2
Rabbit
Antigen Affinity Chromatography
RUO
80328
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BZM5
ULBP2
ULBP2 Fusion Protein Ag3798
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ULBP2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated