Learn More
Bolle Safety Ultim8 Platinum™ Glasses/Goggles
Click to view available options
Lens Tint: Clear
Description
- Pivoting PC/TPR temples
- Sealed TPR foam
- Removable and adjustable strap
- Spectacle/goggle transformation kit
- Platinum™ coating
- Supplied with retainer strap and goggles conversion kit
Specifications
Specifications
Certifications/Compliance | CE, EN166 FT |
Lens Type | Platinum™ |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Glasses/Goggles |
No. per Pack | 1 Ea. |
Weight (Metric) | 64 g |
Lens Material | Polycarbonate |
Videos
-
Bolle Safety ULTIM8
-
Bolle Safety Platinum coating
-
Bolle Safety Platinum efficiency demonstration
Documents
Product Information
Product Information
Bolle Safety ULTIM8
File size: 703 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Bolle Safety Ultim8 Platinum™ Glasses/Goggles >
Spot an opportunity for improvement?Share a Content Correction