Learn More
Thermo Scientific™ Unassembled Convenience Kits: EPA Vials with Caps and Septa
Designed for discrete water sampling applications.
665.00 DKK - 885.00 DKK
Specifications
For Use With (Equipment) Discrete Water Sampling Applications
View More Specs
Products 3
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
12478636
View Documents
Thermo Scientific™
B7920-B
20 mL Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12428646
View Documents
Thermo Scientific™
B7921-B
20 mL Amber This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12438656
View Documents
Thermo Scientific™
B7951-B
40 mL Amber This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Each carton contains shrink-wrapped vials and separately packaged caps and natural-colored PTFE/clear Silicone septa in polybags.
Specifications
Specifications
Discrete Water Sampling Applications