Learn More
Tocris Bioscience™ UNC 0642
Potent and selective G9a and GLP inhibitor
Brand: Tocris Bioscience™ 5132/10
2056.97 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
UNC 0642 is a potent and selective G9a and GLP histone lysine methyltransferase inhibitor (IC50 < 2.5 nM) that exhibits >2,000-fold selectivity for G9a and GLP over PRC2-EZH2 and >20,000-fold selectivity over other methyltransferases. UNC 0642 reduces H3K9 dimethylation levels in MDA-MB-231 cells (IC50 = 110 nM), and displays modest brain penetration in vivo.Specifications
>99% | |
10 mg | |
546.7 |
-20°C | |
UNC 0642 | |
UNC 0642 |
For Research Use Only