Learn More
Tocris Bioscience™ UNC 0642
Potent and selective G9a and GLP inhibitor
Brand:  Tocris Bioscience™ 5132/10
2056.97 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18712449
  • 2605.00 DKK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
UNC 0642 is a potent and selective G9a and GLP histone lysine methyltransferase inhibitor (IC50 < 2.5 nM) that exhibits >2,000-fold selectivity for G9a and GLP over PRC2-EZH2 and >20,000-fold selectivity over other methyltransferases. UNC 0642 reduces H3K9 dimethylation levels in MDA-MB-231 cells (IC50 = 110 nM), and displays modest brain penetration in vivo.
Specifications
Specifications
>99%
10 mg
546.7
-20°C
UNC 0642
UNC 0642
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only