Learn More
Tocris Bioscience™ UNC 1999
Potent and selective EZH2/EZH1 inhibitor
Brand:  Tocris Bioscience™ 4904/50
6387.74 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18790929
  • 8230.00 DKK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
UNC 1999 is a potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.
Negative Control also available.
Specifications
Specifications
>99%
UNC 1999
UNC 1999
50 mg
569.74
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only