Learn More
Köttermann Underbench Chemical Storage Cabinet
This lockable Underbench Chemical Storage Cabinet provides safe storage of aggressive non-flammable chemicals and toxic substances.
Brand: Köttermann 375-0001
This item is not returnable.
View return policy
Description
- For the storage of non-corrosive, non-flammable substances
- Can be integrated in modular cabinet assemblies
- Lockable against unauthorized access
- Capable of connecting to the exhaust air system for permanent ventilation
- Double-walled, noise-insulated doors
Specifications
1 Left-hinged Door, 4 Shelves | |
Hinged, Locking | |
White |
516 x 600 x 1920 mm | |
Non-corrosive, Non-flammable Chemical Storage | |
Steel (Galvanized) |