Learn More
Hughes Unheated Emergency Safety Shower with Eye/Face Wash
Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit. This ANSI compliant model is ideal for use in indoor applications not subject to the possibility of freezing or overheating.
9330.00 DKK - 10920.00 DKK
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
17702348
|
Hughes
HSHD0018GV |
Galvanized Steel |
9330.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
17712348
|
Hughes
HSHD0018SS |
Stainless Steel |
10920.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
Description
- Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit
- ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
- Fitted with foot panel as standard
- Delivers the ANSI recommended 76 liters per minute of potable water from the shower and 12 liters per minute to the eye/face wash
- Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Floor | |
Safety Shower |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hughes Unheated Emergency Safety Shower with Eye/Face Wash