Learn More
OHAUS™ Uni Block
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500588
2760.56 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16670472
  • 2960.00 DKK / Each
Only null left
Estimated Shipment: 29-08-2025
Description
Holds test tubes or vials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 mm Test Tubes
Uni Block
Metal
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Uni Block > 20 mm Test Tubes