Learn More
ABEBA™ Uni6 White Safety Trainers S3
ABEBA™ Uni6 White Safety Trainers S3 are made of white tear and water resistant fabric with a slip resistant PU/TPU outsole.
918.00 DKK
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S3 SRC ESD
Closure Type Laces
Color White
Eyelets Material Steel
Liner Material Breathable silver ions lining
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16367981
View Documents
ABEBA™
31629/35
35
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16377981
View Documents
ABEBA™
31629/36
36
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16387981
View Documents
ABEBA™
31629/37
37
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16397981
View Documents
ABEBA™
31629/38
38
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16307991
View Documents
ABEBA™
31629/39
39
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16317991
View Documents
ABEBA™
31629/40
40
918.00 DKK
1 pair
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
16327991
View Documents
ABEBA™
31629/41
41
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16337991
View Documents
ABEBA™
31629/42
42
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16347991
View Documents
ABEBA™
31629/43
43
918.00 DKK
1 pair
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
16357991
View Documents
ABEBA™
31629/44
44
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16367991
View Documents
ABEBA™
31629/45
45
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16377991
View Documents
ABEBA™
31629/46
46
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16387991
View Documents
ABEBA™
31629/47
47
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
16397991
View Documents
ABEBA™
31629/48
48
918.00 DKK
1 pair
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
Description
Description
  • Comfort, safety and style
  • Comply to EN ISO 20345 : 2012, S3, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Tear and water resistant fabric
  • Steel toe cap (200 joules)
  • Textile antipuncture midsole
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • Removable insole Active comfort
Specifications
Specifications
CE EN ISO 20345:2012 S3 SRC ESD
White
Breathable silver ions lining
Textile
Polyurethane, Thermoplastic Polyurethane
Steel
EN ISO 20345 : 2012; DIN EN 61340
Yes
Laces
Steel
Padded
Slip and abrasion resistant, ESD dissipative
Trainer
1 Pair
Water
Videos
SDS
Documents
Documents
Product Certifications