Learn More
Brady™ Universal Valve Lockout Base and Arm
The Universal Valve Lockout Base and Arm is a modular system which enables you to lock out valves of different types and sizes, offering flexibility and security.
Brand: Brady™ 065401
This item is not returnable.
View return policy
Description
- The Universal Lockout device is a modular system which enables you to lock out valves of different types and sizes.
- No other device offers such flexibility and security.
- Small Dimensions: 22.86 x 157.48 x 142.24 mm (D x W x H) Large Dimensions: 39.37 x 224.03 x 162.05 mm (D x W x H)
Specifications
Large Valve Lockout Base | |
Red | |
1 Ea. |
UNIVERSAL BALL VALVE LOCKOUT DEVICE LRG | |
Large Universal Valve Lockout Base and Arm | |
Large |