missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Stainless Steel Ferrules

Valco/VICI Compatible

424.05 DKK

Specifications

Certifications/Compliance ROHS
Material Stainless Steel
Max. Pressure 20,000 psi
Port Style 10 to 32 coned
Quantity 10/Pk.
View More Specs

Products 1
Product Code Brand For Use With (Equipment) Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Type Price Quantity & Availability  
13080950
missing translation for 'viewDocuments'
Kinesis™
01939-90
0.062 in. OD Tubing Male Hex Nut
424.05 DKK
Pack of 10
In Stock
to see stock.
Add to basket
 
Description

Description

Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar).
Specifications

Specifications

ROHS
20,000 psi
10/Pk.
Stainless Steel
10 to 32 coned
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kinesis™ Stainless Steel Ferrules

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.