missing translation for 'onlineSavingsMsg'
Learn More

RETSCH UR 1 Ultrasonic Bath

22560.00 DKK

Specifications

Timer Yes
Capacity (Metric) 5.7 L
For Use With (Application) Cleaning test sieves and grinding tools quickly and easily
Height (Metric) Exterior 13 cm
Model UR 1
View More Specs

Products 1
Product Code Brand Voltage Price Quantity & Availability  
Product Code Brand Voltage Price Quantity & Availability  
10750093
View Documents
RETSCH
70.791.0001
230 V
22560.00 DKK
Each
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Yes
Cleaning test sieves and grinding tools quickly and easily
UR 1
5.7 L
13 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.