Learn More
WTW™ USB Adapter for WTW™ Meters
The USB adapters ADA USB/Ser and ADA USB are used to connect devices with RS 232 interface
Brand: WTW™ 902881
813.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
WTW offers adapter solutions for mobile energy supply, data transfer of old systems as well as for the connection of conventional pH electrodes to digital systems. The USB adapters ADA USB/Ser and ADA USB are used to connect devices with RS 232 interface to PCs and laptops with USB interfaces, especially all lab and portable meters with RS 232 connector (portable meters 3xxi, benchtop meters inoLab™ Level 2, 730, field meters 197i, 1970i).
Specifications
USB Adapter | |
Laboratory instruments (except OxiCal OC 100/110) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ USB Adapter for WTW™ Meters > For use with: Laboratory instruments (except OxiCal OC 100/110)
Spot an opportunity for improvement?Share a Content Correction