Learn More
Memmert™ Usb User-Id Stick (With User-Id Licence), For Ich750
Memmert™ USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
Brand:  Memmert™ ICH750B33170
Product Code. 16366687
  • 1970.00 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Add to basket
Includes: User-ID License
This item is not returnable. View return policy
Description
Description
  • USB User-ID stick (with User-ID licence)
  • Oven-linked authorisation licence (User-ID-programme) on Memory-stick
  • Prevents undesired manipulation by unauthorised third parties
  • When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Specifications
Only for units with TwinDISPLAY
Units With TwinDISPLAY
USB Stick
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Usb User-Id Stick (With User-Id Licence), For Ich750 >