Learn More
Memmert™ USB User-ID Stick (with User-ID Licence) for SN160plus
USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
Brand: Memmert™ SN160plusB33170
This item is not returnable.
View return policy
Description
- Oven-linked authorization licence (User-ID-program) on Memory-stick
- Prevents undesired manipulation by unauthorised third parties
- When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
SN160plus Sterilizer | |
USB Stick | |
User ID Licence |