Learn More
Memmert™ USB User-ID Stick (with User-ID Licence) for SN160plus
USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
Brand:  Memmert™ SN160plusB33170
Product Code. 17548515
  • 1840.00 DKK / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Description
  • Oven-linked authorization licence (User-ID-program) on Memory-stick
  • Prevents undesired manipulation by unauthorised third parties
  • When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Specifications
SN160plus Sterilizer
USB Stick
User ID Licence