Learn More
Uteroglobin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10490-1-AP-150UL
 View more versions of this product
Product Code. 16856903
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
Uteroglobin encodes a member of the secretoglobin family of small secreted proteins. The encoded protein has been implicated in numerous functions including anti-inflammation, inhibition of phospholipase A2 and the sequestering of hydrophobic ligands. Defects in this gene are associated with a susceptibility to asthma.
Specifications
Specifications
Uteroglobin
Polyclonal
Unconjugated
SCGB1A1
CC10, CC16, CCPBP, CCSP, SCGB1A1, UGB, UP 1, UP1, Urinary protein 1, Urine protein 1, Uteroglobin
Rabbit
Antigen Affinity Chromatography
RUO
22287, 7356
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11684, Q06318
SCGB1A1
Uteroglobin Fusion Protein Ag0758
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Uteroglobin Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated