Learn More
Benchmark Scientific™ UV-C Indicator Card for Research Use Only
UV-C Indicator Card changes color upon exposure to UV-C. For research use only.
Brand:  Benchmark Scientific™ B1450-IC
Product Code. 17994412
  • 908.00 DKK / Pack of 25
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Changes color in the presence of UV-C
  • Used to verify operation of UV-C lamps
  • Double sided
  • For research use only
Specifications
Specifications
Confirm Operation of UV-C Lights
Double-Sided Card
Non-sterile
UV-Clave
Color Change Indicator