Learn More
Benchmark Scientific™ UV-C Transparent Shelf for UV-Clave
UV-C transparent shelf allows for 360 degree irradiation of products in the Benchmark UV-Clave.
Brand:  Benchmark Scientific™ B1450-SH-UV
Product Code. 17914422
  • 1415.00 DKK / Each
Explore more special offers
Description
Description
  • UV-C transparent shelf
  • Proprietary construction
  • Allows for all-around irradiation
  • For use in Benchmark UV-Clave
Specifications
Specifications
Support Items in UV-Clave
UV-Clave
Propretary UV-C Transparent