Learn More
Cytiva UV Flow Cell
Flow cell design with fiber optics ensures high signal-to-noise ratio with minimal drift and refractive index effects.
Brand: Cytiva 18-1111-10
17807.75 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Monitor used as standalone detector or as part of AKTAdesign system
- UNF 10-32 fingertight connectors for capillary tubing with 0.062 in. outer dia.
- Made from PEEK, PTFE, Ti alloy and glass
Specifications
UV Flow Cell | |
Flow cell design with fiber optics ensures high signal-to-noise ratio with minimal drift and refractive index effects | |
PEEK, PTFE, Titanium Alloy, Glass | |
2 mm |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now
Product Content Correction
Product Title
Cytiva UV Flow Cell >
Spot an opportunity for improvement?Share a Content Correction