Learn More
Eppendorf™ UVette™ Adaptors
Brand:  Eppendorf™ 4099005004
589.29 DKK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 10351104
  • 622.48 DKK / Each
Explore more special offers
Specifications
Adaptor
For 20mm H Light Path
Product Title
Eppendorf™ UVette™ Adaptors > For Use With: For 20mm H Light Path