Learn More
uvex i-3 Safety Goggles
Shop All Uvex Products
Click to view available options
Frame Color:
Anthracite Blue
Black, Light Gray
Lens Tint:
Clear
Gray
Description
Specifications
Specifications
Certifications/Compliance | EN 170 |
Lens Type | Uvex™ supravision excellence |
Lens Coating | Anti-fog |
Description | Shield-lens spectacle |
Lens Tint | Clear |
Product Type | Ergonomic Safety Glasses |
Frame Color | Anthracite Blue |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Show More |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
uvex i-3 Safety Goggles >
Spot an opportunity for improvement?Share a Content Correction