Learn More
BrandTech™ VACUUBRAND™ VACUUBUS™ CVC3000 Vacuum Controller Solenoid Valves
For use with CVC3000 Vacuum Controller
2000.00 DKK - 4935.00 DKK
Product Code | Brand | Type | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With | Price | Quantity & Availability | |||||
10681284
|
BrandTech™ VACUUBRAND™
20674291 |
Solenoid Isolation Valve | CVC3000 vacuum controller and DCP3000 vacuum gauge |
4935.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
10353863
|
BrandTech™ VACUUBRAND™
20674220 |
Solenoid Coolant Valve | CVC3000 vacuum controller |
2000.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
10099052
|
BrandTech™ VACUUBRAND™
20674217 |
Solenoid Vent Valve | CVC3000 vacuum controller |
2225.00 DKK
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
|||||
Description
- State-of-the-art vacuum gauges for laboratory and industrial operation have to meet high standards in terms of gauge head resistance to chemicals, reliability in rugged applications and operator-friendly interfaces. VACUUBRAND has a comprehensive range of electronic measuring instruments for the range of atmospheric pressure to 5 x 10-9 mbar.
- by cyclically turning the pump on/off (with CVC 3000 controller with module VMS-B)
- by cyclically opening/closing an in-line solenoid valve (CVC 3000) or
- by changing the pump´s motor speed continuously (VARIO™ pumps in connection with CVC 3000)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BrandTech™ VACUUBRAND™ VACUUBUS™ CVC3000 Vacuum Controller Solenoid Valves