Learn More
Valbenazine, MedChemExpress
MedChemExpress Valbenazine (NBI-98854) is a vesicular monoamine transporter 2 (VMAT2) inhibitor with the Ki of 110-190 nM.
Brand:  Medchem Express HY-16771
Encompass_Preferred
 View more versions of this product
Product Code. 16430286
  • 725.13 DKK / 2mg
Explore more special offers
Description
Description
Valbenazine (NBI-98854) is a vesicular monoamine transporter 2 (VMAT2) inhibitor with the Ki of 110-190 nM.
Specifications
Specifications
Valbenazine
Light Yellow
C24H38N2O4
NBI-98854
O=C([C@@H](N)C(C)C)O[C@@H]1C[C@]2([H])C3=CC(OC)=C(OC)C=C3CCN2C[C@H]1CC(C)C
418.57
Research
Neuroscience-Neuromodulation
1025504-45-3
Research
2 mg
DMSO : 50 mg/mL (119.45 mM; Need ultrasonic)
418.57
98.0%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Valbenazine, MedChemExpress