Learn More
ValueMax™ Extra-Safe™ Lab Coats —
Static-free — no clinging
Brand:  ValueMax™ 23-900-519G
Encompass
 View more versions of this product
Product Code. 12337319
  • POA / Pack of 10
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
This item is not returnable. View return policy
Description
Description
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Color: White
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs
  • Available with knit or traditional collar
  • Knee-length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Specifications
Specifications
General Purpose Lab Coat
Unisex
White
Single-use
3
Yes
3-ply
4X-Large
SMS
Snaps
Knee-length
Knit
3660WH
3-ply SMS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ValueMax™ Extra-Safe™ Lab Coats —