missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP-B Antibody (208), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89626DL594
This item is not returnable.
View return policy
Description
VAP-B Monoclonal specifically detects VAP-B in Human samples. It is validated for Western Blot, ELISA.Specifications
VAP-B | |
Monoclonal | |
DyLight 594 | |
ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human VAP-B (O95292-1; Met 1-Pro 132). | |
0.1 mL | |
Neuroscience | |
9217 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
208 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |