missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAPB Mouse anti-Human, Clone: 4F6A6, Proteintech
Mouse Monoclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | VAPB |
---|---|
Clone | 4F6A6 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827845
|
Proteintech
66191-1-IG-20UL |
20 ÎĽL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16817845
|
Proteintech
66191-1-IG-150UL |
150 ÎĽL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.Specifications
VAPB | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALS8, VAMP associated protein B/C, VAMP B, VAMP B/VAMP C, VAMP C, VAP B, VAP B/VAP C, VAP C, VAPB | |
VAPB | |
IgG1 | |
Protein G | |
Antibody |
4F6A6 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
O95292 | |
9217 | |
VAPB Fusion Protein Ag6313 | |
Primary | |
-20°C | |
VAPB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VAPB Mouse anti-Human, Clone: 4F6A6, Proteintech