Learn More
Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser
Varispenser™ 2, Bottle-Top Dispenser is ideal for dispensing aliquots of liquid from large supply bottles.
2479.88 DKK - 4758.96 DKK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
15707357
|
Eppendorf™
4966000010 |
0.2 to 2 mL |
2605.38 DKK
Each |
In Stock
Log in to see stock. |
||||||
15717357
|
Eppendorf™
4966000029 |
0.5 to 5 mL |
2479.88 DKK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
15727357
|
Eppendorf™
4966000037 |
1 to 10 mL |
2479.88 DKK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
15737357
|
Eppendorf™
4966000045 |
2.5 to 25 mL |
3468.82 DKK
Each |
In Stock
Log in to see stock. |
||||||
15747357
|
Eppendorf™
4966000053 |
5.0 to 50 mL |
3624.44 DKK
Each |
In Stock
Log in to see stock. |
||||||
15757357
|
Eppendorf™
4966000061 |
10 to 100 mL |
4758.96 DKK
Each |
In Stock
Log in to see stock. |
||||||
Description
Varispenser™ 2 is ideal for dispensing aliquots of liquid from large supply bottles.
- Standard GL 45 thread on all sizes for most common bottle threads
- Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
- Screwable air vent cap for easy assembly of drying tube or microfilter
- Oval-shaped dispenser housing for easy transportation
- PFA seal of the sliding piston prevents seizing up
- Fixation slide on interior toothed track for rapid and accurate volume adjustment
- High chemical resistance of all dispensing components
- Telescopic aspirating tubes for variable adjustment to bottle height
- Fully autoclavable without disassembly
- User adjustment option
Specifications
Analog Bottletop Dispenser with Adapters | |
1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser