Learn More
VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26415-1-AP-150UL
 View more versions of this product
Product Code. 16854885
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of this gene are implicated in infantile capillary hemangiomas.
Specifications
Specifications
VEGF Receptor 2
Polyclonal
Unconjugated
KDR
CD309, Fetal liver kinase 1, FLK 1, FLK1, KDR, Kinase insert domain receptor, VEGF Receptor 2, VEGFR, VEGFR 2, VEGFR2
Rabbit
Antigen Affinity Chromatography
RUO
16542, 3791
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P35918, P35968
Kdr
VEGF Receptor 2 Fusion Protein Ag24589
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated