missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26415-1-AP-20UL
This item is not returnable.
View return policy
Description
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of this gene are implicated in infantile capillary hemangiomas.Specifications
VEGF Receptor 2 | |
Polyclonal | |
Unconjugated | |
KDR | |
CD309, Fetal liver kinase 1, FLK 1, FLK1, KDR, Kinase insert domain receptor, VEGF Receptor 2, VEGFR, VEGFR 2, VEGFR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16542, 3791 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35918, P35968 | |
Kdr | |
VEGF Receptor 2 Fusion Protein Ag24589 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction