Learn More
Bochem Evaporating dish Nickel, D=55mm, H=19mm
Brand: Bochem 8040
Evaporating dish Nickel, D=55mm, H=19mm
This item is not returnable.
View return policy
Product Content Correction
Product Title
Bochem Evaporating dish Nickel, D=55mm, H=19mm >
Spot an opportunity for improvement?Share a Content Correction