missing translation for 'onlineSavingsMsg'
Learn More
Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals Product Code.: 11357725

Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11357725
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11357725

Brand: Thermo Scientific Alfa Aesar L02498.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

Solubility
Miscible with tetrahydrofuran, diethyl ether, benzene and dichloromethane.

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and strong alkalis (bases).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.236
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilaneShow MoreShow Less
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C

Specifications

Boiling Point 54°C to 56°C
Melting Point -132°C
Quantity 50 g
UN Number UN1993
Beilstein 956572
Flash Point −24°C (−11°F)
Solubility Information Miscible with tetrahydrofuran,diethyl ether,benzene and dichloromethane.
Formula Weight 100.24
Percent Purity 97%
Odor Mild
Refractive Index 1.391
Density 0.69
Chemical Name or Material Vinyltrimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252361 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberL02498.09,L02498.18,L02498.30
Lot Number10176791 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02498.09,L02498.18,L02498.30
Lot Number10142017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02498.09,L02498.18,L02498.30
Lot Number10235687 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02498.09,L02498.18,L02498.30
Lot Number10149259 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02498.09,L02498.18,L02498.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.