Description
- Sleek, unisex-styled safety glasses
- Comfortable, impact-resistant, lightweight frame
- High-wrap lens fits snug against the face for better coverage
- Integral side shields and browguard
- The modern design offers excellent coverage and field of vision
Specifications
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Spectacles |
No. per Pack | 20 pairs |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ Virtua™ AP Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction