Learn More
Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Brand: Thermo Scientific™ 840-312300
Description
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite Software enhancements include:
Scan:
- Peak pick to identify and label wavelength peaks or valleys
- Overlay spectra for visual comparison
- Import, save or export data in native or CSV format for easy archiving and reporting
- Auto scaling of data plus simple zoom controls for detailed examination of spectra
Rate: For SPECTRONIC 200 and GENESYS 30, measure multi-cell kinetics in parallel or serial mode
- Automatic enzyme activity calculation
- Automatic data storage plus import or export your data in CSV format
- Measure up to 100 samples in a single data set
Fixed:
- Measure at one or multiple wavelengths
- Apply factors and equations to your data to generate results directly
- Addition, subtraction, ratio and baseline correction options
- Display results with units from a menu or define your own custom unit
- Run standard bio-tests using the preprogrammed protein and DNA methods
- Save or print results as a report, or export data in CSV format to use in a template
Quant: More standards and non-linear curve fitting options offer greater confidence and accuracy than can be accomplished under local control on Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers
- Choose from 11 pre-programmed reporting units or define your own
- Apply dilution factors in software to allow you to keep data for all your samples in range without doing off-line calculations
- Automatic data saving for peace of mind
- Save or print your results as a report, or export data in CSV format to use in your own template
Performance Verification:
- Select from the range of available Performance Verification tests that is customized to your instrument’s specifications
- VISIONlite 5 will prompt you to present the correct reference materials in order, and then print a complete report for your compliance file
Accessory control:
Whether your experiment requires automation to enhance sample throughput or thermostatting to preserve precious biological samples, VISIONlite 5 provides full control of the required accessories.
VISIONlite ColocCalc Software:
- Full spectrum color scales and data reporting: Record the required spectra in transmittance or reflectance and calculates color values
- Application-specific color series: Pt-Co/APHA, whiteness, EP color, wine, etc.
- Sample thickness transformation allows for the correction for actual sample thickness and front surface reflection effects based upon a single value, the material's refractive index, or a full surface reflectance spectrum
Mathematical calculations and customization
VISIONlite ColorCalc Software has the ability to perform configurable or custom calculations and to store and present your custom-configured methods on the calculations menu.
Specifications
VISIONlite 5 Software + Cable | |
Thermo Scientific GENESYS 40-180 Spectrophotometers | |
DVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean | |
Interface cable included for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers |