Learn More
Vitronectin Antibody (A18), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-23571AF532
Product Code. 18456215
  • 5380.00 DKK / 0.25mL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Vitronectin Monoclonal antibody specifically detects Vitronectin in Bovine, Equine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
Specifications
Vitronectin
Monoclonal
Alexa Fluor 532
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Bovine, Equine, Rabbit
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
A18
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Lysed bovine corneal endothelial cells and extracellular matrix
0.25 mL
Immunology, Innate Immunity
7448
Store at 4°C in the dark.
IgG1 κ