missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint

Identify media quickly and easily with these color-coded wash bottles. VITsafe™ Narrow-Mouth Wash Bottles with Imprint feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.

Brand:  Vitlab™ 1432829

 View more versions of this product

Product Code. 10644883

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
  • VENT-CAP practically eliminates leakage and prevents almost all static overpressure
  • Color-coded for easy visual identification of the media
Bottles feature durable safety imprint in accordance with Directive (EC) No. 1272/2008 (GHS), as well as with all important information:
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No., hazard pictogram, signal word
  • Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications

Specifications

500 mL
Screw Cap, Spray Tube
12 Pack
Directive (EC) No. 1272/2008 (GHS)
Narrow
Wash Bottle
74 mm
Imprint: Acetone
LDPE
Red
Yes
180 mm
Polypropylene
Vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.