missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ VITsafe™ Wide Mouth Safety Wash Bottles

Product Code. 15722066
500 mL, Tetrahydrofuran (THF), Red
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetic Acid
Hexane
Isopropanol
Methanol
Tetrahydrofuran (THF)
Screw Color:
Green
Red
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Wide Mouth LDPE Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP, easy-fill opening and precise spray control to eliminate drips and leaks and to ensure safe handling. Available for a variety of chemicals.

Working with chemical substances that are sometimes dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.

Constructed of durable, transparent materials
  • Low-density polyethylene (LDPE) bottles
  • Spray tubes made with autoclavable, chemically resistant polypropylene (PP)
  • Wide-mouth bottles for easy and safe filling, even without a funnel
  • Transparent material for easy identification and monitoring
Permanent safety imprint on the bottle with all important information
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No, hazard pictogram, signal word
  • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • In accordance to EC Directive No. 1272/2008 (GHS)
Metal-free, color-colded VENT-CAP
  • Design eliminates almost all static overpressure
  • Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
  • Color-coded caps support easy visual identification of the media
  • Bottles utilize screw cap lids with GL threads for a strong, secure hold
Optimized for precise spray control
  • Finely drawn tip creates a precise spray jet for fine control
  • Optimized medium backflow through the continuous spray tube

Specifications

Capacity (Metric) 500 mL
Color Transparent
Material LDPE
Packaging Individually wrapped in PE bag
Quantity 6Pack
Screw Color Red
Thread Size (Metric) GL 45
Mouth Wide
Height (Metric) 166 mm
Chemical Label Tetrahydrofuran (THF)
Closure Material Polypropylene
Diameter (Metric) 76 mm
Closure Type Vented Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VITLAB™ VITsafe™ Wide Mouth Safety Wash Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.