Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
BIOPHARMA
Brand:  Sartorius™ VN01H22ETO
Product Code. 11776345
  • 5175.00 DKK / Pack of 100
Explore more special offers
Specifications
PCR
30,000 Da
Polycarbonate, Polypropylene
DNA
0.1 to 0.5 mL
Polycarbonate, Polypropylene
Concentration, Desalting, buffer exchange
100/Pk.
Vivacon 500 Centrifugal Unit