Learn More
Sartorius Vivapore 5
Vivapore 5 solvent absorption concentrators are disposable ultrafilters for macromolecule concentration from up to 5 mL aqueous samples.
Brand: Sartorius VP-S005P0008-3
1566.80 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Concentrate equipment-free
- Fill and go
- Tailored process monitoring
- Maintained sample integrity
- Sensitive molecule friendly
Specifications
Ultrafiltration | |
PES | |
5 mL | |
Solvent Absorption Concentrator |
SAN | |
7.5 kDa | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
Sartorius Vivapore 5 > Type:Solvent Absorption
Spot an opportunity for improvement?Share a Content Correction