missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VNN3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-81875
This item is not returnable.
View return policy
Description
VNN3 Polyclonal specifically detects VNN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VNN3 | |
Polyclonal | |
Unconjugated | |
PBS | |
VNN3 | |
VNN3 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human VNN3. The immunogen is located within the last 50 amino acids of VNN3. Amino Acid Squence: RHYEISRDGRLRSRSGAPL | |
Affinity Purified | |
RUO | |
55350 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.5-1 μg/mL, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
HSA238982, MGC171203, vanin 3 | |
Rabbit | |
55 kDa | |
0.1 mg | |
Primary | |
Multiple isoforms of VNN3 are known to exist. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |